Learn More
Biomatik Corporation Mouse Serpin A10 (SERPINA10) ELISA Kit
Supplier:  Biomatik Corporation EKU06955
Encompass_Preferred
Mouse Serpin A10 (SERPINA10) ELISA Kit for detecting SERPINA10 in plasma. Species Reactivity: Mouse (Mus musculus). Detection Range: 6.25-400ng/ml. Sensitivity: 2.74ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8R121. Gene ID: 217847. Target Synonyms: PZI; ZPI; Serine (Cysteine) Proteinase Inhibitor,Clade A (Alpha-1 Antiproteinase,Antitrypsin) Member 10; Protein Z-Related Protease Inhibitor; Protein Z Dependent Protease Inhibitor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6345