Learn More
Biomatik Corporation Rat Podocalyxin (PCX) ELISA Kit, 96 tests
Catalog No. 501503562
Encompass_Preferred
Rat Podocalyxin (PODXL) ELISA Kit for detecting PODXL in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9WTQ2. Gene ID: 192181. Target Synonyms: Gp200; PCLP; PC; PCX; Podocalyxin Like Protein 1; GCTM-2 antigen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3562 Supplier Biomatik Corporation Supplier No. EKU06722
Rat Podocalyxin (PODXL) ELISA Kit for detecting PODXL in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9WTQ2. Gene ID: 192181. Target Synonyms: Gp200; PCLP; PC; PCX; Podocalyxin Like Protein 1; GCTM-2 antigen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.