Learn More
Biomatik Corporation Human Podocin (PDCN) ELISA Kit, 96 tests
Catalog No. 501503563
Encompass_Preferred
The Human Podocin (PDCN) ELISA Kit allows for quantitative determination of Podocin in Tissue homogenates, Urine, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.78-50 ng/mL. Sensitivity: 0.29 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9NP85. Cross Activity: No significant cross-reactivity or interference between Podocin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3563 Supplier Biomatik Corporation Supplier No. EKU06724
The Human Podocin (PDCN) ELISA Kit allows for quantitative determination of Podocin in Tissue homogenates, Urine, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.78-50 ng/mL. Sensitivity: 0.29 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9NP85. Cross Activity: No significant cross-reactivity or interference between Podocin and analogues was observed. Restriction: For Research Use Only.