Learn More
Biomatik Corporation Mouse Secretagogin (SCGN) ELISA Kit
Supplier:  Biomatik Corporation EKU07210
Encompass_Preferred
Mouse Secretagogin (SCGN) ELISA Kit for detecting SCGN in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q91WD9. Gene ID: 214189. Target Synonyms: SECRET; SEGN; CALBL; Calbindin-Like; Secretagogin,EF-Hand Calcium Binding Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6372