Learn More
Biomatik Corporation Rat Podocin (PDCN) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06725
Encompass_Preferred
Rat Podocin (PDCN) ELISA Kit for detecting PDCN in urine, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8K4G9. Target Synonyms: NPHS2; NPHS-2; Nephrosis 2; Idiopathic Steroid-Resistant. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3564