Learn More
Biomatik Corporation Human Poliovirus Receptor (PVR) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06729
Encompass_Preferred
The Human Poliovirus Receptor (PVR) ELISA Kit allows for quantitative determination of Poliovirus Receptor in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1000 pg/mL. Sensitivity: 6.0 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P15151. Cross Activity: No significant cross-reactivity or interference between Poliovirus Receptor and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3567