Learn More
Biomatik Corporation Human Transferrin Receptor 2 (TFR2) ELISA Kit
Supplier:  Biomatik Corporation EKU07791
Encompass_Preferred
Human Transferrin Receptor 2 (TFR2) ELISA Kit for detecting TFR2 in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.263ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9UP52. Gene ID: 7036. Target Synonyms: HFE3; TFRC2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6425