Learn More
Biomatik Corporation Human Polymeric Immunoglobulin Receptor (PIGR) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06742
Encompass_Preferred
The Human Polymeric Immunoglobulin Receptor (PIGR) ELISA Kit allows for quantitative determination of Polymeric Immunoglobulin Receptor in Serum, Plasma, Urine, Saliva and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 62.5-4,000 pg/mL. Sensitivity: 24.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P01833. Cross Activity: No significant cross-reactivity or interference between Polymeric Immunoglobulin Receptor and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3571