Learn More
Biomatik Corporation Mouse Polymeric Immunoglobulin Receptor (PIGR) ELISA Kit, 96 tests
Catalog No. 501503572
Encompass_Preferred
Mouse Polymeric Immunoglobulin Receptor (PIGR) ELISA Kit for detecting PIGR in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.56ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O70570. Gene ID: 18703. Target Synonyms: SC; Secretory Component; Hepatocellular carcinoma-associated protein TB6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3572 Supplier Biomatik Corporation Supplier No. EKU06743
Mouse Polymeric Immunoglobulin Receptor (PIGR) ELISA Kit for detecting PIGR in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.56ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O70570. Gene ID: 18703. Target Synonyms: SC; Secretory Component; Hepatocellular carcinoma-associated protein TB6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.