Learn More
Biomatik Corporation Human Preferentially Expressed Antigen In Melanoma (PRAME) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06749
Encompass_Preferred
The Human Preferentially Expressed Antigen In Melanoma (PRAME) ELISA Kit allows for quantitative determination of Preferentially Expressed Antigen In Melanoma in Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.113 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P78395. Cross Activity: No significant cross-reactivity or interference between Preferentially Expressed Antigen In Melanoma and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3573