Learn More
Biomatik Corporation Human UDP Glycosyltransferase 8 (UGT8) ELISA Kit
Supplier:  Biomatik Corporation EKU08033
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.059ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q16880. Target Synonyms: CGT; Cerebroside synthase; UDP-Galactose Ceramide Galactosyltransferase; 2-Hydroxyacylsphingosine 1-Beta-Galactosyltransferase. Restriction: For Research Use Only.
Catalog No. 50-228-6457