Learn More
Biomatik Corporation Human Pregnancy Associated Plasma Protein A (PAPPA) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06751
Encompass_Preferred
The Human Pregnancy Associated Plasma Protein A (PAPPA) ELISA Kit allows for quantitative determination of Pregnancy Associated Plasma Protein A in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.117 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q13219. Cross Activity: No significant cross-reactivity or interference between Pregnancy Associated Plasma Protein A and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3574