Learn More
Biomatik Corporation Mouse Pregnancy Associated Plasma Protein A (PAPPA) ELISA Kit, 96 tests
Catalog No. 501503575
Encompass_Preferred
Mouse Pregnancy Associated Plasma Protein A (PAPPA) ELISA Kit for detecting PAPPA in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78-5,000pg/ml. Sensitivity: 29pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8R4K8. Gene ID: 18491. Target Synonyms: PAPP-A; PAPA; ASBABP2; DIPLA1; IGFBP-4ase; PAPPA1; Pappalysin 1; Insulin-Like Growth Factor Binding Protein-4 Protease; Differentially Placenta 1 Expressed Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3575 Supplier Biomatik Corporation Supplier No. EKU06752
Mouse Pregnancy Associated Plasma Protein A (PAPPA) ELISA Kit for detecting PAPPA in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78-5,000pg/ml. Sensitivity: 29pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8R4K8. Gene ID: 18491. Target Synonyms: PAPP-A; PAPA; ASBABP2; DIPLA1; IGFBP-4ase; PAPPA1; Pappalysin 1; Insulin-Like Growth Factor Binding Protein-4 Protease; Differentially Placenta 1 Expressed Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.