Learn More
Biomatik Corporation Mouse Procalcitonin (PCT) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06766
Encompass_Preferred
Mouse Procalcitonin (PCT) ELISA Kit for detecting PCT in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10pg/ml. Sensitivity: 0.069pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P70160. Gene ID: 12310. Target Synonyms: Pro-Calcitonin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3579