Learn More
Biomatik Corporation Rat Procalcitonin (PCT) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06767
Encompass_Preferred
Rat Procalcitonin (PCT) ELISA Kit for detecting PCT in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P01257. Gene ID: 24241. Target Synonyms: Pro-Calcitonin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3580