Learn More
Biomatik Corporation Mouse Procollagen I C-Terminal Propeptide (PICP) ELISA Kit, 96 tests
Catalog No. 501503584
Encompass_Preferred
Mouse Procollagen I C-Terminal Propeptide (PICP) ELISA Kit for detecting PICP in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 125-8,000pg/ml. Sensitivity: 50pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P11087. Gene ID: 12842. Target Synonyms: P1CP; C-Propeptide Of Type I Procollagen; Procollagen I Carboxy Terminal Propeptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3584 Supplier Biomatik Corporation Supplier No. EKU06771
Mouse Procollagen I C-Terminal Propeptide (PICP) ELISA Kit for detecting PICP in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 125-8,000pg/ml. Sensitivity: 50pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P11087. Gene ID: 12842. Target Synonyms: P1CP; C-Propeptide Of Type I Procollagen; Procollagen I Carboxy Terminal Propeptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.