Learn More
Biomatik Corporation Mouse Procollagen I N-Terminal Propeptide (PINP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06774
Encompass_Preferred
Mouse Procollagen I N-Terminal Propeptide (PINP) ELISA Kit for detecting PINP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 46.88-3,000pg/ml. Sensitivity: 17.71pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P11087. Gene ID: 12842. Target Synonyms: P1NP; N-Propeptide Of Type I Procollagen; Procollagen I Amino Terminal Propeptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3586