Learn More
Biomatik Corporation Human Xanthine Dehydrogenase (XDH) CLIA Kit
Supplier:  Biomatik Corporation EKU10007
Encompass_Preferred
Human Xanthine Dehydrogenase (XDH) CLIA Kit for detecting XDH in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 13.7-10,000pg/ml. Sensitivity: 4.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P47989. Gene ID: 7498. Target Synonyms: XO; XOR; XOD; Xanthine Oxidase; NAD+-Xanthine Dehydrogenase; Xanthine-NAD+ Oxidoreductase; Xanthine/NAD+ Oxidoreductase; Xanthine Oxidoreductase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6639