Learn More
Biomatik Corporation Human Activated Protein C (APC) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU10263
Encompass_Preferred
Human Activated Protein C (APC) High-sensitive ELISA Kit for detecting APC in plasma. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.6pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P04070. Gene ID: 5624. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6682