Learn More
Biomatik Corporation Estradiol (E2) Instant ELISA Kit
Supplier:  Biomatik Corporation EKU10337
Encompass_Preferred
Estradiol (E2) Instant ELISA Kit for detecting E2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Universal. Detection Range: 12.35-1000pg/ml. Sensitivity: 4.65pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 1h, 10min. Target Synonyms: 17B-Estradiol; Oestradiol; Beta-Estradiol. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6698