Learn More
Biomatik Corporation Human Progestagen Associated Endometrial Protein (PAEP) ELISA Kit, 96 tests
Catalog No. 501503598
Encompass_Preferred
The Human Progestagen Associated Endometrial Protein (PAEP) ELISA Kit allows for quantitative determination of Progestagen Associated Endometrial Protein in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 4.69-300 ng/mL. Sensitivity: 2.1 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P09466. Cross Activity: No significant cross-reactivity or interference between Progestagen Associated Endometrial Protein and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3598 Supplier Biomatik Corporation Supplier No. EKU06797
The Human Progestagen Associated Endometrial Protein (PAEP) ELISA Kit allows for quantitative determination of Progestagen Associated Endometrial Protein in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 4.69-300 ng/mL. Sensitivity: 2.1 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P09466. Cross Activity: No significant cross-reactivity or interference between Progestagen Associated Endometrial Protein and analogues was observed. Restriction: For Research Use Only.