Learn More
Biomatik Corporation Anandamide (AEA) ELISA Kit
Supplier:  Biomatik Corporation EKU10634
Encompass_Preferred
Anandamide (AEA) ELISA Kit for detecting AEA in serum, plasma, other biological fluids. Species Reactivity: Universal. Detection Range: 2.47-200ng/ml. Sensitivity: 0.89ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: N-Arachidonoylethanolamine. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6724