Learn More
Biomatik Corporation Mouse Programmed Cell Death Protein 1 Ligand 1 (PDL1) ELISA Kit, 96 tests
Catalog No. 501503603
Encompass_Preferred
Mouse Programmed Cell Death Protein 1 Ligand 1 (PDL1) ELISA Kit for detecting PDL1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9EP73. Gene ID: 60533. Target Synonyms: CD274; PDCD1LG1; B7-H; B7H1; PD-L1; PDCD1L1; PDCD1-LG1; Programed Death Ligand 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3603 Supplier Biomatik Corporation Supplier No. EKU06803
Mouse Programmed Cell Death Protein 1 Ligand 1 (PDL1) ELISA Kit for detecting PDL1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9EP73. Gene ID: 60533. Target Synonyms: CD274; PDCD1LG1; B7-H; B7H1; PD-L1; PDCD1L1; PDCD1-LG1; Programed Death Ligand 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.