Learn More
Biomatik Corporation Human Proteasome Subunit Beta Type 10 (PSMb10) ELISA Kit
Supplier:  Biomatik Corporation EKU11373
Encompass_Preferred
Human Proteasome Subunit Beta Type 10 (PSMb10) ELISA Kit for detecting PSMb10 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.117ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P40306. Gene ID: 5699. Target Synonyms: LMP10; MECL1; Low molecular mass protein 10; Macropain subunit MECl-1; Multicatalytic endopeptidase complex subunit MECl-1; Proteasome subunit beta-2i. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6908