Learn More
Biomatik Corporation Mouse Von Willebrand Factor (vWF) ELISA Kit
Supplier:  Biomatik Corporation EKU11530
Encompass_Preferred
Mouse Von Willebrand Factor (vWF) ELISA Kit for detecting vWF in plasma. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.12-200ng/ml. Sensitivity: 1.22ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8CIZ8. Gene ID: 22371. Target Synonyms: F8VWF; VWD; von Willebrand antigen 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6944