Learn More
Biomatik Corporation Human Proopiomelanocortin (POMC) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06841
Encompass_Preferred
The Human Proopiomelanocortin (POMC) ELISA Kit allows for quantitative determination of Proopiomelanocortin in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 123.5-10,000 pg/mL. Sensitivity: 43.9 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P01189. Cross Activity: No significant cross-reactivity or interference between Proopiomelanocortin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3614