Learn More
Biomatik Corporation Mouse Proprotein Convertase Subtilisin/Kexin Type 9 (PCSK9) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06848
Encompass_Preferred
Mouse Proprotein Convertase Subtilisin/Kexin Type 9 (PCSK9) ELISA Kit for detecting PCSK9 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1000pg/ml. Sensitivity: 6.0pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q80W65. Gene ID: 100102. Target Synonyms: FH3; HCHOLA3; NARC1; Hypercholesterolemia,Autosomal Dominant 3; Neural apoptosis-regulated convertase 1; Proprotein convertase 9; Subtilisin/kexin-like protease PC9. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3618