Learn More
Biomatik Corporation Rat Proprotein Convertase Subtilisin/Kexin Type 9 (PCSK9) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06849
Encompass_Preferred
Rat Proprotein Convertase Subtilisin/Kexin Type 9 (PCSK9) ELISA Kit for detecting PCSK9 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 3.12-200ng/ml. Sensitivity: 1.21ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P59996. Gene ID: 298296. Target Synonyms: FH3; HCHOLA3; NARC1; Hypercholesterolemia,Autosomal Dominant 3; Neural apoptosis-regulated convertase 1; Proprotein convertase 9; Subtilisin/kexin-like protease PC9. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3619