Learn More
Biomatik Corporation Chicken Fatty Acid Binding Protein 2, Intestinal (FABP2) ELISA Kit
Supplier:  Biomatik Corporation EKU11907
Encompass_Preferred
Chicken Fatty Acid Binding Protein 2, Intestinal (FABP2) ELISA Kit for detecting FABP2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.117ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q7ZZZ5. Gene ID: 422678. Target Synonyms: FABPI; IFABP; I-FABP; Intestinal-type fatty acid-binding protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7023