Learn More
Biomatik Corporation Human Prostaglandin D2 Synthase, Brain (PGD2S) ELISA Kit, 96 tests
Catalog No. 501503621
Encompass_Preferred
The Human Prostaglandin D2 Synthase, Brain (PGD2S) ELISA Kit allows for quantitative determination of Prostaglandin D2 Synthase, Brain in Serum, Plasma, cerebrospinal fluid and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.129 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P41222. Cross Activity: No significant cross-reactivity or interference between Prostaglandin D2 Synthase, Brain and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3621 Supplier Biomatik Corporation Supplier No. EKU06853
The Human Prostaglandin D2 Synthase, Brain (PGD2S) ELISA Kit allows for quantitative determination of Prostaglandin D2 Synthase, Brain in Serum, Plasma, cerebrospinal fluid and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.129 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P41222. Cross Activity: No significant cross-reactivity or interference between Prostaglandin D2 Synthase, Brain and analogues was observed. Restriction: For Research Use Only.