Learn More
Biomatik Corporation Prostaglandin E2 (PGE2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06858
Encompass_Preferred
Prostaglandin E2 (PGE2) ELISA Kit for detecting PGE2 in serum, plasma, other biological fluids. Species Reactivity: Universal. Detection Range: 24.69-2,000pg/ml. Sensitivity: 8.67pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: PG-E2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3624