Learn More
Biomatik Corporation Human Prostaglandin Endoperoxide Synthase 1 (PTGS1) ELISA Kit, 96 tests
Catalog No. 501503625
Encompass_Preferred
The Human Prostaglandin Endoperoxide Synthase 1 (PTGS1) ELISA Kit allows for quantitative determination of Prostaglandin Endoperoxide Synthase 1 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.625-40 ng/mL. Sensitivity: 0.225 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P23219. Cross Activity: No significant cross-reactivity or interference between Prostaglandin Endoperoxide Synthase 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3625 Supplier Biomatik Corporation Supplier No. EKU06859
The Human Prostaglandin Endoperoxide Synthase 1 (PTGS1) ELISA Kit allows for quantitative determination of Prostaglandin Endoperoxide Synthase 1 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.625-40 ng/mL. Sensitivity: 0.225 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P23219. Cross Activity: No significant cross-reactivity or interference between Prostaglandin Endoperoxide Synthase 1 and analogues was observed. Restriction: For Research Use Only.