Learn More
Biomatik Corporation Rat Prostaglandin Endoperoxide Synthase 1 (PTGS1) ELISA Kit, 96 tests
Catalog No. 501503626
Encompass_Preferred
Rat Cyclooxygenase 1 (COX-1) ELISA Kit for detecting COX-1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.245ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q63921. Target Synonyms: PTGS1; COX3; PCOX1; PGG/HS; PGHS1; PHS1; PTGHS; Prostaglandin Endoperoxide Synthase 1; Prostaglandin H2 Synthase 1; Prostaglandin G/H Synthase And Cyclooxygenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3626 Supplier Biomatik Corporation Supplier No. EKU06860
Rat Cyclooxygenase 1 (COX-1) ELISA Kit for detecting COX-1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.245ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q63921. Target Synonyms: PTGS1; COX3; PCOX1; PGG/HS; PGHS1; PHS1; PTGHS; Prostaglandin Endoperoxide Synthase 1; Prostaglandin H2 Synthase 1; Prostaglandin G/H Synthase And Cyclooxygenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.