Learn More
Biomatik Corporation Human Protease Activated Receptor 1 (PAR1) ELISA Kit, 96 tests
Catalog No. 501503629
Encompass_Preferred
The Human Protease Activated Receptor 1 (PAR1) ELISA Kit allows for quantitative determination of Protease Activated Receptor 1 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.054 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P25116. Cross Activity: No significant cross-reactivity or interference between Protease Activated Receptor 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3629 Supplier Biomatik Corporation Supplier No. EKU06869
The Human Protease Activated Receptor 1 (PAR1) ELISA Kit allows for quantitative determination of Protease Activated Receptor 1 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.054 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P25116. Cross Activity: No significant cross-reactivity or interference between Protease Activated Receptor 1 and analogues was observed. Restriction: For Research Use Only.