Learn More
Biomatik Corporation Mouse Phosphatidylserine (PS) ELISA Kit
Supplier:  Biomatik Corporation EKU12289
Encompass_Preferred
Mouse Phosphatidylserine (PS) ELISA Kit for detecting PS in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 12.35-1,000ng/ml. Sensitivity: 5.79ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: Ptd-L-Ser; PtdSer. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7113