Learn More
Biomatik Corporation Human Protease, Serine 1 (PRSS1) ELISA Kit, 96 tests
Catalog No. 501503631
Encompass_Preferred
The Human Protease, Serine 1 (PRSS1) ELISA Kit allows for quantitative determination of Protease, Serine 1 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1,000 pg/mL. Sensitivity: 6.9 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P07477. Cross Activity: No significant cross-reactivity or interference between Protease, Serine 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3631 Supplier Biomatik Corporation Supplier No. EKU06876
The Human Protease, Serine 1 (PRSS1) ELISA Kit allows for quantitative determination of Protease, Serine 1 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1,000 pg/mL. Sensitivity: 6.9 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P07477. Cross Activity: No significant cross-reactivity or interference between Protease, Serine 1 and analogues was observed. Restriction: For Research Use Only.