Learn More
Biomatik Corporation Human Protectin (CD59) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06889
Encompass_Preferred
The Human Protectin (CD59) ELISA Kit allows for quantitative determination of Protectin in Serum, Plasma, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 12.5-800 pg/mL. Sensitivity: 5.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P13987. Cross Activity: No significant cross-reactivity or interference between Protectin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3632