Learn More
Biomatik Corporation Mouse Protein C (PROC) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06893
Encompass_Preferred
Mouse Protein C (PROC) ELISA Kit for detecting PROC in plasma. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P33587. Gene ID: 19123. Target Synonyms: PRO-C; VKDPC; PC; Vitamin K Dependent Protein C; Inactivator Of Coagulation Factors Va And VIIIa; Autoprothrombin IIA; Blood coagulation factor XIV; Activation peptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3634