Learn More
Biomatik Corporation Rat Protein C (PROC) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06894
Encompass_Preferred
Rat Protein C (PROC) ELISA Kit for detecting PROC in plasma. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.067ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P31394. Gene ID: 25268. Target Synonyms: PRO-C; VKDPC; PC; Vitamin K Dependent Protein C; Inactivator Of Coagulation Factors Va And VIIIa; Autoprothrombin IIA; Blood coagulation factor XIV; Activation peptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3635