Learn More
Biomatik Corporation Rat Low Density Lipoprotein cholesterol (LDL-C) ELISA kit
Supplier:  Biomatik Corporation EKC31524
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 125 ng/mL-8000 ng/mL. Sensitivity: 62.5 ng/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-228-7211