Learn More
Biomatik Corporation Human Protein Disulfide Isomerase A4 (PDIA4) ELISA Kit, 96 tests
Catalog No. 501503640
Encompass_Preferred
The Human Protein Disulfide Isomerase A4 (PDIA4) ELISA Kit allows for quantitative determination of Protein Disulfide Isomerase A4 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.61 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P13667. Cross Activity: No significant cross-reactivity or interference between Protein Disulfide Isomerase A4 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3640 Supplier Biomatik Corporation Supplier No. EKU06903
The Human Protein Disulfide Isomerase A4 (PDIA4) ELISA Kit allows for quantitative determination of Protein Disulfide Isomerase A4 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.61 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P13667. Cross Activity: No significant cross-reactivity or interference between Protein Disulfide Isomerase A4 and analogues was observed. Restriction: For Research Use Only.