Learn More
Biomatik Corporation Bovine Leptin, Lep Elisa Kit
Supplier:  Biomatik Corporation EKC31714
Encompass_Preferred
Bovine Leptin, Lep Elisa Kit for detecting Leptin in serum, plasma, tissue homogenates. Species Reactivity: Bovine (Bos taurus). Detection Range: 3.12-50ng/ml. Sensitivity: 3.12ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P50595; LEP. Target Synonyms: LEP; OB; OBS; Leptin; Obesity factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7221