Learn More
Biomatik Corporation Mouse Protein Kinase, DNA Activated, Catalytic Polypeptide (PRKDC) ELISA Kit, 96 tests
Catalog No. 501503645
Encompass_Preferred
Mouse Protein Kinase, DNA Activated, Catalytic Polypeptide (PRKDC) ELISA Kit for detecting PRKDC in saliva, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.118ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P97313. Gene ID: 19090. Target Synonyms: HYRC; HYRC1; DNPK1; p350; DNAPK; XRCC7; DNA-PKcs. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3645 Supplier Biomatik Corporation Supplier No. EKU06934
Mouse Protein Kinase, DNA Activated, Catalytic Polypeptide (PRKDC) ELISA Kit for detecting PRKDC in saliva, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.118ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P97313. Gene ID: 19090. Target Synonyms: HYRC; HYRC1; DNPK1; p350; DNAPK; XRCC7; DNA-PKcs. Restrictions: For Research Use Only. Not for use in diagnostic procedures.