Learn More
Biomatik Corporation Human Protein L-Isoaspartate-O-Methyltransferase (PCMT1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06936
Encompass_Preferred
The Human Protein L-Isoaspartate-O-Methyltransferase (PCMT1) ELISA Kit allows for quantitative determination of Protein L-Isoaspartate-O-Methyltransferase in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 31.2-2000 pg/mL. Sensitivity: 13.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P22061. Cross Activity: No significant cross-reactivity or interference between Protein L-Isoaspartate-O-Methyltransferase and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3646