Learn More
Biomatik Corporation Canine Matrix Metalloproteinase 13, Mmp-13 Elisa Kit
Supplier:  Biomatik Corporation EKC31866
Encompass_Preferred
Canine Matrix Metalloproteinase 13, Mmp-13 Elisa Kit for detecting Matrix Metalloproteinase 13, Mmp-13 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: F1PVS9. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7233