Learn More
Biomatik Corporation Human Protein Z Dependent Protease Inhibitor (ZPI) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06954
Encompass_Preferred
The Human Protein Z Dependent Protease Inhibitor (ZPI) ELISA Kit allows for quantitative determination of Protein Z Dependent Protease Inhibitor in Plasma. Species Reactivity: Homo sapiens (Human). Detection Range: 1.25-80 ng/mL. Sensitivity: 0.49 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9UK55. Cross Activity: No significant cross-reactivity or interference between Protein Z Dependent Protease Inhibitor and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3650