Learn More
Biomatik Corporation Human Protein Zero, Myelin (MPZ) ELISA Kit, 96 tests
Catalog No. 501503651
Encompass_Preferred
The Human Protein Zero, Myelin (MPZ) ELISA Kit allows for quantitative determination of Protein Zero, Myelin in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.114 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P25189. Cross Activity: No significant cross-reactivity or interference between Protein Zero, Myelin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3651 Supplier Biomatik Corporation Supplier No. EKU06956
The Human Protein Zero, Myelin (MPZ) ELISA Kit allows for quantitative determination of Protein Zero, Myelin in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.114 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P25189. Cross Activity: No significant cross-reactivity or interference between Protein Zero, Myelin and analogues was observed. Restriction: For Research Use Only.