Learn More
Biomatik Corporation Chicken Parathyroid Hormone, Pth Elisa Kit
Supplier:  Biomatik Corporation EKC31976
Encompass_Preferred
Chicken Parathyroid Hormone, Pth Elisa Kit for detecting Parathyroid Hormone in serum, plasma, tissue homogenates. Species Reactivity: Chicken (Gallus). Detection Range: 6.25-100pg/ml. Sensitivity: 6.25pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P15743; PTH. Target Synonyms: PTH; Parathyroid hormone; PTH. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7247