Learn More
Biomatik Corporation Human Proteoglycan 4 (PRG4) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06961
Encompass_Preferred
The Human Proteoglycan 4 (PRG4) ELISA Kit allows for quantitative determination of Proteoglycan 4 in Serum, Plasma, synovial fluid and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 62.5-4000 pg/mL. Sensitivity: 25.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q92954. Cross Activity: No significant cross-reactivity or interference between Proteoglycan 4 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3654