Learn More
Biomatik Corporation Fish Catalase (CAT) ELISA kit
Supplier:  Biomatik Corporation EKC32145
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Fish. Detection Range: 125 mU/mL-2000 mU/mL. Sensitivity: 62.5 mU/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: H2L8K1. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-228-7260